Skip to Content
Merck

63301-011

Sigma-Aldrich

PCRboost®

1 mL bottle, for improved PCR performance

Synonym(s):

PCRboost®

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352200
NACRES:
NA.52
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

liquid

usage

28 reactions

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

PCRboost® is a unique reagent that enhances end-point and reverse transcription-PCR performance by improving sensitivity and specificity during amplification of genomic DNA or RNA templates. To use, simply replace the water in your PCR with PCRboost®. That′s all. It is designed to enhance the amplification of DNA without the need for time consuming optimizations. PCRboost® successfully amplifies thermally degraded DNA resulting in a visible amplicon that can be used for downstream applications such as cloning and sequencing.

Don′t see what you are looking for? Custom stabilization services for molecular diagnostic assay and reagent manufacturers are available through Sigma′s partner, Biomatrica. PCRstable® enables the preservation of PCR-based assay components in any format (liquid or dry).

Features and Benefits

Boost PCR results for:

  • Degraded FFPE samples and bisulphite-treated samples (methylation studies)
  • Multiplex PCR
  • Low-copy transcripts
  • Samples containing inhibitory factors (i.e. humic acid, heme, and indigo dye)

No changes to your PCR protocol. Simply replace the water in 
your reaction.

PCR products are compatible with all downstream applications.

Packaging

1 bottle

Physical form

1.0 mL PCRboost® in a 1.5 mL screw-top conical tube

Legal Information

PCRboost is a registered trademark of Biomatrica
PCRstable is a registered trademark of Biomatrica

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service