Merck
HomeProtein Labeling & ModificationGlycosyltransferases: Tools for Synthesis and Modification of Glycans
Glycosyltransferases: Tools for Synthesis and Modification of Glycans
The presence of multiple functional groups and stereocenters in complex carbohydrates makes them challenging targets for the organic chemist. Chemical synthesis research has not yielded robust, automated protocols comparable to those developed for the preparation of peptides and oligonucleotides. There are two major obstacles to the large-scale, chemical synthesis of carbohydrates and glycoconjugates: 1-5
  • Multiple hydroxyl groups with similar reactivities must be differentiated in order to create the desired regioselective and stereospecific glycosidic bonds. Laborious manipulation of protecting groups and complex synthetic schemes are required to prevent reactions with undesired hydroxyl sites. The large number of potential linkages between specific monosaccharide units requires effective regioselective and stereospecific activation of either glycosyl donors or acceptors.
  • As many carbohydrates are only soluble in water, synthetic manipulation requires either an adaptation of organic reactions to aqueous media or a reversible modification of the carbohydrates to achieve solubility in non-aqueous solvents.
Glycosyltransferases from the Leloir pathway 6-8 have been proven to be viable alternatives to chemical synthesis in the preparation of oligosaccharides. 1,2,9-13 As more of these transferases are isolated from natural sources or produced by recombinant technology, chemists have recognized enzymatic glycosylation as the preferred method to complement classical synthetic techniques. Leloir glycosyltransferases are highly regioselective and stereospecific with respect to the glycosidic linkages formed. They incorporate unprotected sugar precursors, avoid tedious chemical modifications, and provide oligosaccharides in high yields.
The biosynthesis of oligosaccharides, catalyzed by glycosyltransferases from the Leloir pathway, resembles the corresponding chemical procedure (Figure 1). A donor sugar is activated in the first step, followed by the transfer of the activated sugar to an appropriate acceptor sugar. Leloir glycosyltransferases primarily utilize one of eight different nucleotide mono- or diphosphates (UDP-Glc, UDP-GlcNAc, UDP-Gal, UDP-GalNAc, GDP-Man, GDPFuc, UDP-GlcA, and CMP-NeuAc) as monosaccharide donors to build a new glycosidic bond. 7
Enzymatic-catalyzed glycosylation
Figure 1. Enzymatic-catalyzed glycosylation using β(1→4)Galactosyltransferase [β(1→4) GalT].
Glycosyltransferases are specific for the type of linkage (α or β), and the linkage position of the glycoside bond formed [e.g. α(1→3) or β(1→4)]. Glycosyltransferases were initially considered to be specific for a single glycosyl donor and acceptor, which led to the “one enzyme–one linkage” concept. 28,29 Subsequent observations have refuted the theory of absolute enzymatic specificity by describing the transfer of analogs of some nucleoside mono- or diphosphate sugar donors. 30-36 Glycosyltransferases can tolerate modifications to the acceptor sugar, as long as the acceptor meets specific structural requirements (e.g. appropriate stereochemistry and availability of the reactive hydroxyl group involved in the glycosidic bond).
A major limitation to enzyme-catalyzed glycosylation reactions is the glycosyltransferase inhibition caused by nucleoside diphosphates generated during the reaction. Two strategies have been identified to prevent enzymatic inhibition (Figure 2):
1. Phosphatase is added to the reaction to degrade the nucleoside diphosphates by removal of the phosphate group (Figure 2A).23
2. Nucleoside diphosphates are recycled to the appropriate nucleotide diphosphates by employing multi-enzyme regeneration schemes. Although several different enzymes and cofactors are involved in these in situ regeneration schemes, the method avoids the use of stoichiometric amounts of sugar nucleotides (Figure 2B).24-26
Methods for avoiding enzyme inhibition in glycosyltransferasecatalyzed synthesis
Figure 2. Methods for avoiding enzyme inhibition in glycosyltransferasecatalyzed synthesis: (A) Addition of phosphatase. (B) Recycling of sugar nucleotides (NDP = nucleoside diphosphates, NTP = nucleoside triphosphates, N = nucleoside, Pi = phosphate).
In contrast to organic chemical synthesis, enzymatic glycosylation has potential for application use within biological systems, where the modification of glycosylation sites may be used to investigate the regulation of cell signalling processes. Various application strategies for glycosyltransferases have employed an assortment of glycosyl donors and reaction conditions for the synthesis of carbohydrates and the glycosylation of natural products.27,28
α(1→3)Galactosyltransferase
α(1→3)Galactosyltransferase (EC 2.4.1.151; α(1→3)GalT) is responsible for the formation of α-galactosyl epitopes bearing α-Gal(1→3)-β-Gal-OR termini. The interaction of α-Gal epitopes (Galili antigens) on the surface of animal cells (e.g. porcine endothelial cells) with anti-galactosyl antibodies present in human serum is believed to be the main cause in antibodymediated hyperacute rejection following xenotransplantation.43-51 Experimental attempts to overcome hyperacute rejection revealed the need for α-Gal oligo saccharides, synthetic α-Gal analogs, and mimetics with high affinity to anti-Gal antibodies. Earlier methods to chemically synthesize α-Gal trisaccharides were tedious,49-51 while glycosidase-catalyzed trans glycosyl ation reactions to form the desired α-Gal(1→3)-β-Gal-OR linkage resulted in poor yields and regioselectivities.38-40 Using recombinant α(1→3) galactosyltransferase, α-Gal epitopes and several derivatives have been synthesized on a preparative scale.41
α(1→3)Galactosyltransferase transfers a galactose unit from the activated donor UDP-galactose (UDP-Gal) to the 3-hydroxy site of a terminal β-linked galactose, resulting in an α-linkage. Several studies of α(1→3)galactosyl transferase substrate specificity have been carried out which show a high acceptor promiscuity of the enzyme in vitro.38-40 Acceptors that have been successfully used include lactose, β-lactosyl azide, β-thiophenyl lactoside, N-acetyllactosamine derivatives, lactosamine,41 and a wide range of N-acylderivatives of type II disaccharides. Carbamate groups, protected amino acid residues, lipophilic, and hydrophilic aromatic residues can replace the natural occurring N-acetyl group.6 α(1→3)Galactosyl transferase can transfer galactose to an unnatural hindered tertiary hydroxyl group of the acceptor sugar, yielding an acetal formation reaction with a highly deactivated hydroxyl group that is extremely difficult to synthesize by chemical methods.42
β(1→4)Galactosyltransferase
The synthesis and substrate specificity of β(1→4)Galactosyltransferase (EC 2.4.1.22; β(1→4)GalT) from bovine milk has been extensively investigated.2,9-12,43-49 β(1→4)GalT catalyzes the transfer of galactose from UDP-galactose (UDP-Gal) to the 4-hydroxy site of N-acetyl-D-glucosamine (GlcNAc) and β-linked GlcNAc subunits to yield β-lactosamine (β-LacNAc) and β-Gal(1→4)-β-GlcNAc structures respectively.50 Both α- and β-glycosides of glucose have been used as acceptors by β(1→4)GalT, with α-glucosides requiring the presence of α-lactalbumin.26 The enzyme forms a heterodimeric complex with α-lactalbumin, altering the specificity so that D-glucose becomes the preferred acceptor. Thus, addition of α-lactalbumin promotes the formation of lactose (β-Gal(1→4)- Glc-OH). Numerous other acceptor substrates for the β(1→4)GalTcatalyzed transfer of galactose have been described, including 2-deoxy glucose, D-xylose, 5-thioglucose, N-acetylmuramic acid, and myo-inositol. 6-O-Fucosylated and sialyated modifications may also serve as acceptors,51 as well as 3-O-methyl-GlcNAc,24 3-deoxy- GlcNAc, 3-O-allyl-GlcNAc-β-OBu and 3-oxo-GlcNAc.66 Several modifications of GlcNAc that have been employed as acceptor substrates are illustrated (Figure 3).9
Modifications of GlcNAc employed as acceptors in β(1→4)GalT catalyzed transfer of galactose.
Figure 3. Modifications of GlcNAc employed as acceptors in β(1→4)GalT catalyzed transfer of galactose.
β(1→4)GalT cannot utilize D-mannose, D-allose, D-galactose, or D-ribose as substrates.11-12 Monosaccharides displaying a negative charge, such as glucuronic acid and α-glucose 1-phosphate, are also not tolerated as substrates. Azasugars and glucals have been shown to be very weak acceptors.24 Modified nucleotide sugar donor substrates have a slower rate of enzyme-catalyzed transfer.11,12
N-Acetylglucosaminyl amino acids and peptides have been successfully galactosylated to produce glycopeptides with a disaccharide moiety. Subsequent extension of the carbohydrate chain was accomplished by employing α(2→6)sialyltransferase.53-54
An asparagine-bound trisaccharide was prepared using combined chemo-enzymatic synthesis.53 Attachment of galactose to a N-acetylglucosaminyl oligopeptide was followed by sialylation with α(2→3)sialyltransferase and fucosylation with α(2→3)- fucosyltransferase, which yielded a glycopeptide containing a tetrasaccharide moiety.55
Since different glycosides of N-acetylglucosamine and glucose can be used as acceptors in β(1→4)GalT-catalyzed galactose transfer, the enzymatic method has been used to modify pharmacologically interesting glycosides.56-59 β(1→4)GalT has been used to attach galactose to the bioactive glycosides elymoclavine-17-O-β-Dglucopyranoside, 56 stevioside and steviolbioside,60 colchicoside and fraxin,61 and different ginsenosides.62 Conjugation of galactose with glycosides demonstrates the potential application in drug delivery by increasing the solubility and bioavailability of large hydrophobic molecules under mild conditions. C-Glycoside analogs of the naturally occurring glycopeptide linkages (N-acetylglucosamine β-linked to either asparagine or serine) generated high yields of the corresponding C-lactosides.63
β(1→4)GalT has been employed in solid-phase oligosaccharide synthesis on polymer supports such as polyacrylamide or water-soluble poly(vinyl alcohol). The resulting galactosylated oligosaccharides are cleaved from the polymers photochemically or with chymotrypsin.64
α-1,3-Fucosyltransferase VI
α(1→3)Fucosyltransferase (α(1→3)FucT) catalyzes the transfer of L-fucose from the donor guanosine diphosphate-β-L-fucose (GDP-Fuc) to the free 3-hydroxy position as an α-orientation65,66 and tolerates a wide range of acceptors (Figure 4).67
Minimum structural requirements for an acceptor employed
Figure 4. Minimum structural requirements for an acceptor employed in α(1→3)FucT catalyzed transfer of L-fucose include: 6’-OH, free 3-OH, β(1→4)-linkage and 2-NH-acylation. An acyl lactosamine is shown as an example acceptor.
The number and linkage type of fucose residues in N-glycans and the fucosylation pattern varies with the organism, the tissue, and the developmental and physiological status of the cell. 68 Fucose is normally attached:
• To a N-glycan by α(1→2)-linkage to galactose (Gal)
• To a N-glycan by α(1→3), α(1→4), or α(1→6)-linkage to an N-acetylglucosamine (GlcNAc) residue
• To a peptide by direct O-linkage to serine/threonine
The terminal step in the biosynthetic pathway of fucosecontaining saccharides is the transfer of L-fucose from GDPFuc to the corresponding glycoconjugate acceptor catalyzed by fucosyltransferase. 67-71 Fucosylated glycan structures within glycopeptides, glycoproteins and glycolipids play a central role in cell-cell interactions and cell migration, increasing the significance of the study of fucosyltransferase expression, inhibition and regulation. More than 150 complete or partial sequences of fucosyltransferases can be found through protein sequence databases such as Swiss Institute of Bioinformatics Swiss-Prot system www.expasy.ch.
Glycosyltransferase Kits
As part of our commitment to biotransformation technologies, we have developed recombinant glycosyltransferases and kits for preparative carbohydrate synthesis and directed modification of carbohydrate moieties. The enzymatic synthesis reactions go to completion rapidly and specifically, eliminating the need to isolate the desired glycan from closely related by-products.
Our glycosyltransferase kits contain the enzyme, the appropriate nucleotide sugar donor, and all other components required for the transfer of a specific mono saccharide moiety to an acceptor substrate on a small preparative scale. Our glycosylation kits include alkaline phosphatase to degrade nucleotide diphosphate and prevent the inhibition of glycosyl transferase activity.
• Unique glycosyltransferases – deliver regiospecific and stereospecific glycosylation
• Individual enzyme aliquots for each glycosylation reaction – prevent enzyme activity loss and cross-contamination
Glycosyltransferases and nucleotide sugar donors are available separately*.
* Sales restrictions may apply. Please contact your local sales representative.
Materials
Loading
References
1.
Toone EJ, Simon ES, Bednarski MD, Whitesides GM. 1989. Enzyme-catalyzed synthesis of carbohydrates. Tetrahedron. 45(17):5365-5422. https://doi.org/10.1016/s0040-4020(01)89487-4
2.
Koeller KM, Wong C. 2000. Synthesis of Complex Carbohydrates and Glycoconjugates:  Enzyme-Based and Programmable One-Pot Strategies. Chem. Rev.. 100(12):4465-4494. https://doi.org/10.1021/cr990297n
3.
Paulsen H. 1982. Advances in Selective Chemical Syntheses of Complex Oligosaccharides. Angew. Chem. Int. Ed. Engl.. 21(3):155-173. https://doi.org/10.1002/anie.198201553
4.
Paulsen H. 1984. Haworth Memorial Lecture. Synthesis of complex oligosaccharide chains of glycoproteins. Chem. Soc. Rev.. 13(1):15. https://doi.org/10.1039/cs9841300015
5.
Kunz H. 1987. Synthesis of Glycopeptides, Partial Structures of Biological Recognition Components[New Synthetic Methods(67)]. Angew. Chem. Int. Ed. Engl.. 26(4):294-308. https://doi.org/10.1002/anie.198702941
6.
Leloir LF. 1971. Two Decades of Research on Biosynthesis of Saccharides. Science. 172(3990):1299-1303. https://doi.org/10.1126/science.172.3990.1299
7.
Kornfeld R, Kornfeld S. 1985. Assembly of Asparagine-Linked Oligosaccharides. Annu. Rev. Biochem.. 54(1):631-664. https://doi.org/10.1146/annurev.bi.54.070185.003215
8.
Watkins WM. 1986. Glycosyltransferases, early history, development and future prospects. Carbohydrate Research. 149(1):1-12. https://doi.org/10.1016/s0008-6215(00)90364-1
9.
Drueckhammer DG, Hennen WJ, Pederson RL, Barbas, Iii CF, Gautheron CM, Krach T, Wong C. 1991. Enzyme Catalysis in Synthetic Carbohydrate Chemistry. Synthesis. 1991(07):499-525. https://doi.org/10.1055/s-1991-26506
10.
Wong C, Halcomb RL, Ichikawa Y, Kajimoto T. 1995. Enzyme in der organischen Synthese: das Problem der molekularen Erkennung von Kohlenhydraten (Teil 2). Angew. Chem.. 107(5):569-593. https://doi.org/10.1002/ange.19951070505
11.
Drauz K, Waldmann H. 1995. Enzyme Catalysis in Organic Synthesis, . 279. VCH, Weinheim,:
12.
Wong C, Whitesides G. 1994. Enzymes in Synthetic Organic Chemistry, Tetrahedron Organic Chemistry Series, . Vol. 12. Oxford: Elsevier Science Ltd.
13.
Wang P. 2000. et al., Curr. Opin. Drug Discov. Devel.,. 3, 756,.
14.
Beyer K. 1981. Neuere Aspekte der Pentazocin-Analytik.52-57. https://doi.org/10.1007/978-3-642-68081-6_7
15.
Evan Sadler J, Beyer TA, Oppenheimer CL, Paulson JC, Prieels J, Rearick JI, Hill RL. 1982. [41] Purification of mammalian glycosyltransferases.458-514. https://doi.org/10.1016/0076-6879(82)83043-7
16.
Morin MJ, Porter CW, Petrie CR, Korytnyk W, Bernacki RJ. 1983. Effects of a membrane sugar analogue, 6-deoxy-6-fluoro-d-galactose, on the L1210 leukemic cell ectosialyltransferase system. Biochemical Pharmacology. 32(3):553-561. https://doi.org/10.1016/0006-2952(83)90538-5
17.
McDowell W, Grier TJ, Rasmussen JR, Schwarz RT. 1987. The role of C-4-substituted mannose analogues in protein glycosylation. Effect of the guanosine diphosphate esters of 4-deoxy-4-fluoro-d-mannose and 4-deoxy-d-mannose on lipid-linked oligosaccharide assembly. 248(2):523-531. https://doi.org/10.1042/bj2480523
18.
Shibaev VN. 1978. Biosynthesis of Salmonella O-antigenic polysaccharides: specificity of glycosyl transferases. 50(11-12):1421-1436. https://doi.org/10.1351/pac197850111421
19.
Higa H, Paulson J. 1985. Sialylation of glycoprotein oligosaccharides with N-acetyl-, N-glycolyl-, and NO-diacetylneuraminic acids. Journal of Biological Chemistry.. 260(15) 8838-49.
20.
Conradt H, Bünsch A, Brossmer R. 1984. Preparation of 9-fluoro-9-deoxy-N-[2-14C]acetylneuraminic acid. 170(2):295-300. https://doi.org/10.1016/0014-5793(84)81331-9
21.
GROSS HJ, BUNSCH A, PAULSON JC, BROSSMER R. 1987. Activation and transfer of novel synthetic 9-substituted sialic acids. Eur J Biochem. 168(3):595-602. https://doi.org/10.1111/j.1432-1033.1987.tb13458.x
22.
Augé C, Gautheron C. 1988. An efficient synthesis of cytidine monophospho-sialic acids with four immobilized enzymes.. Tetrahedron Letters. 29(7):789-790. https://doi.org/10.1016/s0040-4039(00)80210-5
23.
Unverzagt C, Kunz H, Paulson JC. 1990. High-efficiency synthesis of sialyloligosaccharides and sialoglycopeptides. J. Am. Chem. Soc.. 112(25):9308-9309. https://doi.org/10.1021/ja00181a037
24.
Augé C, David S, Mathieu C, Gautheron C. 1984. Synthesis with immobilized enzymes of two trisaccharides, one of them active as the determinant of a stage antigen.. Tetrahedron Letters. 25(14):1467-1470. https://doi.org/10.1016/s0040-4039(01)80188-x
25.
Ichikawa Y, Wang R, Wong C. 1994. [7] Regeneration of sugar nucleotide for enzymatic oligosaccharide synthesis.107-127. https://doi.org/10.1016/s0076-6879(94)47009-x
26.
Ichikawa M, Schnaar RL, Ichikawa Y. 1995. Application of sucrose phosphorylase reaction in one-pot enzymatic galactosylation: Scavenger of phosphate and generation of glucose 1-phosphate in situ. Tetrahedron Letters. 36(48):8731-8732. https://doi.org/10.1016/0040-4039(95)01896-p
27.
Guo Z, Wang PG. 1997. Utilization of glycosyltransferases to change oligosaccharide structures. Appl Biochem Biotechnol. 68(1-2):1-20. https://doi.org/10.1007/bf02785977
28.
Riva S. 2001. Biocatalytic modification of natural products. Current Opinion in Chemical Biology. 5(2):106-111. https://doi.org/10.1016/s1367-5931(00)00178-2
29.
Galili U. 1993. Interaction of the natural anti-Gal antibody with ?-galactosyl epitopes: a major obstacle for xenotransplantation in humans. Immunology Today. 14(10):480-482. https://doi.org/10.1016/0167-5699(93)90261-i
30.
Galili U. 1993. Evolution and pathophysiology of the human natural anti-?-galactosyl IgG (anti-Gal) antibody. Springer Semin Immunopathol. 15(2-3): https://doi.org/10.1007/bf00201098
31.
Gustafsson K, Strahan K, Preece A. 1994. alpha1,3Galactosyltransferase: A Target for in vivo Genetic Manipulation in Xenotransplantation. Immunol Rev. 141(1):59-70. https://doi.org/10.1111/j.1600-065x.1994.tb00872.x
32.
Sandrin MS, Vaughan HA, McKenzie IF. 1994. Identification of Gal(?1,3)Gal as the major epitope for pig-to-human vascularised xenografts. Transplantation Reviews. 8(3):134-149. https://doi.org/10.1016/s0955-470x(05)80033-3
33.
Sandrin M, Mckenzie IFC. 1994. Galalpha(1,3)Gal, the Major Xenoantigen(s) Recognised in Pigs by Human Natural Antibodies. Immunol Rev. 141(1):169-190. https://doi.org/10.1111/j.1600-065x.1994.tb00877.x
34.
Cooper DKC, Koren E, Oriol R. 1994. Oligosaccharides and Discordant Xenotransplantation. Immunol Rev. 141(1):31-58. https://doi.org/10.1111/j.1600-065x.1994.tb00871.x
35.
Jacquinet J, Duchet D, Milat M, Sinay P. 1981. Synthesis of blood-group substances. Part 11. Synthesis of the trisaccharide O-α-D-galactopyranosyl-(1→ 3)-O-β-D-galactopyranosyl-(1→ 4)-2-acetamido-2-deoxy-D-glucopyranose.. Journal of the Chemical Society, Perkin Transactions 1..326-30.
36.
Matsuzaki Y, Ito Y, Nakahara Y, Ogawa T. 1993. Synthesis of branched poly-N-acetyl-lactosamine type pentaantennary pentacosasaccharide: Glycan part of a glycosyl ceramide from rabbit erythrocyte membrane. Tetrahedron Letters. 34(6):1061-1064. https://doi.org/10.1016/s0040-4039(00)77492-2
37.
Reddy GV, Jain RK, Bhatti BS, Matta KL. 1994. Synthesis of ?-d-galactopyranosyl-linked oligosaccharides containing the ?-Gal ? ?-Gal ? GlcNAc sequence employing methyl-2,3,4,6-tetra-O-(4-methoxybenzyl)-1-thio- ?-d-galactopyranoside as an efficient glycosyl donor. Carbohydrate Research. 263(1):67-77. https://doi.org/10.1016/0008-6215(94)00153-7
38.
Nilsson KG. 1997. Glycosidase-catalysed synthesis of di- and trisaccharide derivatives related to antigens involved in the hyperacure rejection of xenotransplants. Tetrahedron Letters. 38(1):133-136. https://doi.org/10.1016/s0040-4039(96)02235-6
39.
Vic G. 1997. J. Chem. Soc. Chem Commun..1169.
40.
Matsuo I, Fujimoto H, Isomura M, Ajisaka K. 1997. Chemoenzymatic synthesis of Gal?1-3Gal, Gal?1-3Gal?1-4GlcNAc and their PEG-conjugates.. Bioorganic & Medicinal Chemistry Letters. 7(3):255-258. https://doi.org/10.1016/s0960-894x(97)00017-6
41.
Fang J, Li J, Chen X, Zhang Y, Wang J, Guo Z, Zhang W, Yu L, Brew K, Wang PG. 1998. Highly Efficient Chemoenzymatic Synthesis of ?-Galactosyl Epitopes with a Recombinant ?(1?3)-Galactosyltransferase. J. Am. Chem. Soc.. 120(27):6635-6638. https://doi.org/10.1021/ja9808898
42.
Qian X, Sujino K, Otter A, Palcic MM, Hindsgaul O. 1999. Chemoenzymatic Synthesis of ?-(1?3)-Gal(NAc)-Terminating Glycosides of Complex Tertiary Sugar Alcohols. J. Am. Chem. Soc.. 121(51):12063-12072. https://doi.org/10.1021/ja993004g
43.
Schanbacher F, Ebner K. 1970. Galactosyltransferase acceptor specificity of the lactose synthetase A protein. Journal of Biological Chemistry.. 245(19) 5057-61.
44.
Berliner L, Davis M, Ebner K, Beyer T, Bell J. 1984. The lactose synthase acceptor site: a structural map derived from acceptor studies. Mol Cell Biochem. 62(1): https://doi.org/10.1007/bf00230075
45.
Nunez HA, Barker R. 1980. Enzymic synthesis and carbon-13 nuclear magnetic resonance conformational studies of disaccharides containing .beta.-D-galactopyranosyl and .beta.-D-[1-13C]galactopyranosyl residues. Biochemistry. 19(3):489-495. https://doi.org/10.1021/bi00544a015
46.
Trayer I, Hill R. 1971. The purification and properties of the A protein of lactose synthetase. Journal of Biological Chemistry.. 246(21) 6666-75.
47.
Andrews P. 1970. Purification of lactose synthetase a protein from human milk and demonstration of its interaction with ?-lactalbumin. 9(5):297-300. https://doi.org/10.1016/0014-5793(70)80382-9
48.
Barker R, Olsen K, Shaper J, Hill R. 1972. Agarose derivatives of uridine diphosphate and N-acetylglucosamine for the purification of a galactosyltransferase. Journal of biological Chemistry.. 247(22) 7135-47.
49.
Rao AK, Garver F, Mendicino J. 1976. Biosynthesis of the carbohydrate units of immunoglobulins. 1. Purification and properties of galactosyltransferases from swine mesentary lymph nodes. Biochemistry. 15(23):5001-5009. https://doi.org/10.1021/bi00668a009
50.
Baisch G, Öhrlein R, Ernst B. 1996. Enzymatic galactosylation of non-natural glucosamide-acceptors. Bioorganic & Medicinal Chemistry Letters. 6(7):749-754. https://doi.org/10.1016/0960-894x(96)00117-5
51.
Palcic MM, Srivastava OP, Hindsgaul O. 1987. Transfer of d-galactosyl groups to 6-O-substituted 2-acetamido-2-deoxy-d-glucose residues by use of bovine d-galactosyltransferase. Carbohydrate Research. 159(2):315-324. https://doi.org/10.1016/s0008-6215(00)90224-6
52.
Wong CH, Ichikawa Y, Krach T, Gautheron-Le Narvor C, Dumas DP, Look GC. 1991. Probing the acceptor specificity of .beta.-1,4-galactosyltransferase for the development of enzymatic synthesis of novel oligosaccharides. J. Am. Chem. Soc.. 113(21):8137-8145. https://doi.org/10.1021/ja00021a045
53.
Thiem J, Wiemann T. 1990. Kombinierte chemoenzymatische Synthese vonN-Glycoproteinbausteinen. Angew. Chem.. 102(1):78-80. https://doi.org/10.1002/ange.19901020119
54.
Baisch G, Öhrlein R. 1996. Chemo-enzymatische Synthese von Sialyl-Lewisx-Glycopeptiden. Angew. Chem.. 108(16):1949-1952. https://doi.org/10.1002/ange.19961081617
55.
Unverzagt C, Kunz H, Paulson JC. 1990. High-efficiency synthesis of sialyloligosaccharides and sialoglycopeptides. J. Am. Chem. Soc.. 112(25):9308-9309. https://doi.org/10.1021/ja00181a037
56.
K?en V, Augé C, Sedmera P, Havlí?ek V. ?-Glucosyl and ?-galactosyl transfer catalysed by ?-1,4-galactosyltransferase in preparation of glycosylated alkaloids. J. Chem. Soc., Perkin Trans. 1.(17):2481-2484. https://doi.org/10.1039/p19940002481
57.
Riva S. 2002. Enzymatic modification of the sugar moieties of natural glycosides. Journal of Molecular Catalysis B: Enzymatic. 19-2043-54. https://doi.org/10.1016/s1381-1177(02)00150-9
58.
RIVA S, MONTI D, LUISETTI M, DANIELI B. 1998. Enzymatic Modification of Natural Compounds with Pharmacological Propertiesa. Annals NY Acad Sci. 864(1 ENZYME ENGINE):70-80. https://doi.org/10.1111/j.1749-6632.1998.tb10289.x
59.
Panza L, Chiappini PL, Russo G, Monti D, Riva S. 1997. Stereoselective enzymatic galactosylation of C-glucosides. J. Chem. Soc., Perkin Trans. 1.(9):1255-1256. https://doi.org/10.1039/a701747b
60.
Danieli B, Luisetti M, Schubert-Zsilavecz M, Likussar W, Steurer S, Riva S, Monti D, Reiner J. 1997. Regioselective Enzyme-Mediated Glycosylation of Natural Polyhydroxy Compounds. Part 1. Galactosylation of stevioside and steviolbioside. Helv. Chim. Acta. 80(4):1153-1160. https://doi.org/10.1002/hlca.19970800412
61.
Riva S, Sennino B, Zambianchi F, Danieli B, Panza L. 1997. Effect of organic cosolvents on the stability and activity of the ?-1,4-galactosyltransferase from bovine colostrum. Carbohydrate Research. 305(3-4):525-531. https://doi.org/10.1016/s0008-6215(97)00273-5
62.
Gebhardt S, Bihler S, Schubert‐Zsilavecz M, Riva S, Monti D, Falcone L, Danieli B. 2002. Biocatalytic Generation of Molecular Diversity: Modification of Ginsenoside Rb1 by β‐1, 4‐Galactosyltransferase and Candida antarctica Lipase, Part 4. Helvetica chimica acta. 85(7) 1943-59.
63.
Tarantini L, Monti D, Panza L, Prosperi D, Riva S. 2001. Enzymatic galactosylation of C-glycosides analogues en route to C-glycopeptides. Journal of Molecular Catalysis B: Enzymatic. 11(4-6):343-348. https://doi.org/10.1016/s1381-1177(00)00020-5
64.
Zehavi U, Herchman M. 1984. Enzymic synthesis of oligosaccharides on an ?-chymotrypsin-sensitive polymer. O-(?-d-Galactopyranosyl)-(1?4)-O-(?-d-glucopyranosyl)-(1?4)-d-glucopyranose. Carbohydrate Research. 133(2):339-342. https://doi.org/10.1016/0008-6215(84)85212-x
65.
Weston B, Smith P, Kelly R, Lowe J. 1992. Molecular cloning of a fourth member of a human alpha (1, 3) fucosyltransferase gene family. Multiple homologous sequences that determine expression of the Lewis x, sialyl Lewis x, and difucosyl sialyl Lewis x epitopes. Journal of Biological Chemistry. 267(34) 24575-84.
66.
Weston B. 1993. J. Biol. Chem. 268, 18398 .
67.
Baisch G, Öhrlein R, Katopodis A, Ernst B. 1996. Enzymatic fucosylation of non-natural trisaccharides with cloned fucosyltransferase VI. Bioorganic & Medicinal Chemistry Letters. 6(7):759-762. https://doi.org/10.1016/0960-894x(96)00119-9
68.
Staudacher E. 1999. Fucose in N-glycans: from plant to man. 1473(1):216-236. https://doi.org/10.1016/s0304-4165(99)00181-6
69.
Javaud C. 2003. 118(2/3):157-170. https://doi.org/10.1023/a:1024101625214
70.
de Vries T, Knegtel RM, Holmes EH, Macher BA. 2001. Fucosyltransferases: structure/function studies. Glycobiology. 11(10):119R-128R. https://doi.org/10.1093/glycob/11.10.119r
71.
Paschinger K, Staudacher E, Stemmer U, Fabini G, Wilson IBH. 2005. Fucosyltransferase substrate specificity and the order of fucosylation in invertebrates. 15(5):463-474. https://doi.org/10.1093/glycob/cwi028