Skip to Content
Merck

SRE0098

Sigma-Aldrich

Bovine Serum Albumin

≥98%, New Zealand origin, ≤3 EU/mg endotoxin, western blot: suitable, electrophoresis: suitable, ELISA: suitable

Synonym(s):

Bovine Serum Albumin, Albumin bovine serum, BSA, Bovine albumin

Sign Into View Organizational & Contract Pricing

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bovine Serum Albumin, heat shock fraction, New Zealand origin, protease free, IgG free, fatty acid free, Ph 7.0, ≥98%

Assay

≥98%

form

lyophilized powder

mol wt

~66 kDa
~66 kDa

purified by

heat shock fractionation

origin

New Zealand origin

free fatty acid content content

≤0.05% (NEFA)

impurities

essentially globulin free
≤3 EU/mg endotoxin

ash

<2%

loss

≤5%

pH

7.0

solubility

water: soluble (40 mg/ml)

cation traces

heavy metals (as Pb): ≤10 ppm

foreign activity

protease ≤0.005 U/mg

storage temp.

2-8°C

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library