Skip to Content
Merck

SCT017

Sigma-Aldrich

LuminiCell Tracker NIR- Cell Labeling Kit

new

Synonym(s):

Live Cell Fluorescent Tracker, Live Cell Imaging Probe, NIR Live Cell Tracker, Near infrared fluorescent cell tracker

Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41106514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Level

packaging

kit of 1 kit

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable

shipped in

2-8°C

storage temp.

2-8°C

Target description

Long-term noninvasive cell tracking by fluorescent probes and quantum dots is increasingly useful to life scientists and biomedical engineers. Current methods used to fluorescently tag cells have been limited by short signal duration, high background autofluorescence or complex and time-consuming molecular cloning manipulations using fluorescent proteins such as GFP.

LuminiCell Tracker are biocompatible organic fluorescent nanoparticles based on Aggregation Induced Emission (AIEdot) technology. Aggregation-induced emission (AIE) molecules emit fluorescence differently than other popular fluorophores such as fluorescent proteins: propeller-shaped AIE fluorogens are non-emissive in solutions but become highly fluorescent upon aggregate formation. As a consequence, LuminiCell Tracker have very high fluorescence intensities with minimal signal quenching, allowing live cell fluorescent tagging for up to 10 days in vitro and 21 days in vivo. Such properties make these unique trackers optimal candidates for long interval live cell bioimaging experiments.

Storage and Stability

Store at 2 - 8°C, protected from light. Spin vial briefly to collect contents at bottom of vial before opening. Do not freeze.

Other Notes

SCT017 contains highly emissive fluorescent organic nanoparticles, designed for rapid labeling of live cells with biocompatibility. These particles can be readily taken up by diverse cell types, often within one hour. Once within cells, these particles demonstrate bright fluorescence with remarkable photostability. and emit durable signal for multiple cell generations with negligible cytotoxicity. Labelled cells can be monitored using a variety of fluorescence techniques such as confocal microscopy, multiphoton microscopy and flow cytometry.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library