Skip to Content
Merck

546488

Sigma-Aldrich

2,5-Bis(chloromethyl)-1,4-bis(3′,7′-dimethyloctyloxy)benzene

98%

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
(ClCH2)2C6H2(O(CH2)2CH(CH3)(CH2)3CH(CH3)2)2
CAS Number:
Molecular Weight:
487.59
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Assay

98%

mp

60-63 °C (lit.)

SMILES string

CC(C)CCCC(C)CCOc1cc(CCl)c(OCCC(C)CCCC(C)C)cc1CCl

Application

Conducting polymer precursor for synthesis of substituted PPV.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library