Skip to Content
Merck

540765

Sigma-Aldrich

Poly(p-xylene tetrahydrothiophenium chloride) solution

0.25 wt. % in H2O

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
(C12H15ClS)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


concentration

0.25 wt. % in H2O

refractive index

n20/D 1.3347

pH

1.67

density

1.01 g/mL at 25 °C

fluorescence

λex 322 nm; λem 432 nm in H2O

storage temp.

2-8°C

General description

This intermediate polyelectrolyte can be cast into films of any dimension by evaporation or spin-coating. Thermal curing (250-300°C) under vacuum to drive off the remaining tetrahydrothiophene, HCl, and water yields unsubstituted PPV.

Application

Light-emitting polymer (PPV) precursor.


Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library