MilliporeSigma
Phénols
Phénols application for SPME, suitable for GC
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne capillaire SLB®-5ms pour GC
L × I.D. 30 m × 0.25 mm, df 0.50 μm
Fibre SPME
Réf. du produit
Description
Tarif
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating
needle size 23 ga, df 60 μm(PEG, for use with autosampler
CONDITIONS
sample/matrix
Phenols at 25 ppb in 0.1M phosphate buffer, pH 2, with 25% NaCl, 9 mL in 10 mL screw cap vial
SPME fiber
60 μm Carbowax (PEG) (57354-U)
extraction
30 min direct immersion with agitation using CombiPal autosampler
desorption process
(30 min. direct immersion with agitation using CombiPal autosampler)
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.5μm film
oven
50°C(2.0 min) to 160°C @ 10°C/min to 280°C @ 20°C/min
inj. temp.
Splitless/split with 0.75mm ID liner and Merlin Microseal
detector
ITMS m/z=45-275 @ 0.7 μsec per scan, transfer line 310°C
carrier gas
Helium, 1mL/min constant flow, 8.7 psi
Description
Informations légales
Carbowax is a registered trademark of Union Carbide
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany