MilliporeSigma
Composés volatiles dans le vin blanc
Composés volatiles dans le vin blanc suitable for GC, application for SPME
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne capillaire pour la GC VOCOL®
L × I.D. 30 m × 0.25 mm, df 1.50 μm
Composant de phase mobile
Réf. du produit
Description
Tarif
Methyl formate
analytical standard
Fibre SPME
Réf. du produit
Description
Tarif
Unité de fibres SPME, carboxène/polydiméthylsiloxane (CAR/PDMS)
df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Produit(s) apparenté(s)
Réf. du produit
Description
Tarif
Support de fibres SPME
for use with Manual Sampling
Support de fibres SPME
for use with CTC CombiPAL, Gerstel MPS2
Étalon
Réf. du produit
Description
Tarif
Hexyl acetate
analytical standard
Octanoic acid
analytical standard
Ethyl butyrate
analytical standard
Isoamyl alcohol
≥98%, FG
2-Methyl-1-butanol
≥99%
Sulfur dioxide
≥99.9%
Ethyl hexanoate
analytical standard
Ethyl octanoate
analytical standard
2,3-Butanediol
analytical standard, mixture of racemic and meso forms
CONDITIONS
sample/matrix
white wine + 25% NaCl
SPME fiber
Carboxen/PDMS, 75 μm (57318)
extraction
10 min, 40 °C (headspace)
desorption process
3 min at 290 °C
column
VOCOL, 30 m × 0.25 mm I.D., 1.5 μm (24205-U)
oven
40 °C (2 min) to 220 °C at 10 °C/min
inj. temp.
290 °C
detector
GC-MS, quadrupole, m/z = 31-240
carrier gas
helium, 35 cm/sec
injection
splitless/split, closed 1 min
liner
0.75 mm liner
sample
white wine
Description
Remarque sur l'analyse
In this application, SPME was used for the analysis of volatiles in white wine using a headspace method. The presence of aroma compounds in white wine is one of the most important attributes of its final quality. A Carboxen/PDMS fiber was inserted into the headspace of the vial for 10 minutes at 40 °C and analyzed by GC/MS. The thick film of the VOCOL GC column helps focus the volatile analytes.
Informations légales
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany