MilliporeSigma
  • Involvement of cyclic nucleotide-gated channels in spontaneous activity generated in isolated interstitial cells of Cajal from the rabbit urethra.
Involvement of cyclic nucleotide-gated channels in spontaneous activity generated in isolated interstitial cells of Cajal from the rabbit urethra.
European journal of pharmacology (2017-08-22)
Maria Sancho, Eamonn Bradley, Angeles Garcia-Pascual, Domingo Triguero, Keith D Thornbury, Mark A Hollywood, Gerard P Sergeant
RÉSUMÉ
Cyclic nucleotide-gated (CNG) channels are non-selective cation channels that mediate influx of extracellular Na