- Frequency of CD4+CD161+ T Cell and Interleukin-10 Expression in Inflammatory Bowel Diseases.
Frequency of CD4+CD161+ T Cell and Interleukin-10 Expression in Inflammatory Bowel Diseases.
Acta histochemica et cytochemica (2017-04-08)
Kazuyo Tsuchiya, Takuto Ikeda, Baatarsuren Batmunkh, Narantsog Choijookhuu, Hidenobu Ishizaki, Masayuki Hotokezaka, Yoshitaka Hishikawa, Atsushi Nanashima
PMID28386147
RÉSUMÉ
Mucosal immune dysregulation associated with T cells plays a critical role in the development of inflammatory bowel diseases (IBD). However, the definite significances of these cells in IBD still remain unclear. Therefore, we investigated the population and expression of CD4