MilliporeSigma
Prognostic value of increased expression of RACO-1 in patients with hepatitis B-related hepatocellular carcinoma.
Therapeutics and clinical risk management (2017-03-01)
Jian-Yao Chen, Li-Ping Liu, Jiang-Feng Xu
RÉSUMÉ
RING domain AP-1 coactivator-1 (RACO-1) is a coactivator that links c-Jun to growth factor signaling and is essential for AP-1 function. This study aimed to investigate the expression and clinical significance of RACO-1 protein in hepatitis B virus (HBV)-related hepatocellular carcinoma (HCC) in China. A total of 136 tissue samples of HBV-related HCC were detected by immunohistochemistry (including 76 patients in training cohort and 60 patients in validation cohort). Correlation between RACO-1 expression and clinicopathologic features of HBV-related HCC was analyzed in both the cohorts. RACO-1 expression was significantly higher in HBV-related HCC tissues than in adjacent non-tumor liver tissues. All the patients were divided into two groups: the low expression group and the high expression group. RACO-1 expression was significantly related to vascular invasion (