MilliporeSigma
Phase I study of RGB-286638, a novel, multitargeted cyclin-dependent kinase inhibitor in patients with solid tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-07-16)
Diane A J van der Biessen, Herman Burger, Peter de Bruijn, Cor H J Lamers, Nicole Naus, Hannes Loferer, Erik A C Wiemer, Ron H J Mathijssen, Maja J A de Jonge
RÉSUMÉ
RGB-286638 is a multitargeted inhibitor with targets comprising the family of cyclin-dependent kinases (CDK) and a range of other cancer-relevant tyrosine and serine/threonine kinases. The objectives of this first in human trial of RGB-286638, given i.v. on days 1 to 5 every 28 days, were to determine the maximum tolerated dose (MTD) and to evaluate the pharmacokinetic (PK) and pharmacodynamic (PD) profiles of this new drug. Sequential cohorts of 3 to 6 patients were treated per dose level. Blood, urine samples, and skin biopsies for full PK and/or PD analyses were collected. Twenty-six patients were enrolled in 6-dose levels from 10 to 160 mg/d. Four dose-limiting toxicities were observed in 2 of the 6 patients enrolled at the highest dose level. These toxicities were AST/ALT elevations in 1 patient, paroxysmal supraventricular tachycardias (SVTs), hypotension, and an increase in troponin T in another patient. The plasma PK of RGB-286638 was shown to be linear over the studied doses. The interpatient variability in clearance was moderate (variation coefficient 7%-36%). The PD analyses in peripheral blood mononuclear cells, serum (apoptosis induction) and skin biopsies (Rb, p-Rb, Ki-67, and p27(KIP1) expression) did not demonstrate a consistent modulation of mechanism-related biomarkers with the exception of lowered Ki-67 levels at the MTD level. The recommended MTD for phase II studies is 120 mg/d. RGB-286638 is tolerated when administered at 120 mg/d for 5 days every 28 days. Prolonged disease stabilization (range, 2-14 months) was seen across different dose levels.