MilliporeSigma
  • Absence of gamma-interferon-inducible lysosomal thiol reductase (GILT) is associated with poor disease-free survival in breast cancer patients.
Absence of gamma-interferon-inducible lysosomal thiol reductase (GILT) is associated with poor disease-free survival in breast cancer patients.
PloS one (2014-10-22)
Yu-Juan Xiang, Ming-Ming Guo, Cheng-Jun Zhou, Lu Liu, Bo Han, Ling-Yu Kong, Zhong-Cheng Gao, Zhong-Bing Ma, Lu Wang, Man Feng, Hai-Ying Chen, Guo-Tao Jia, De-Zong Gao, Qiang Zhang, Liang Li, Yu-Yang Li, Zhi-Gang Yu
RÉSUMÉ
Tumor immunosurveillance is known to be of critical importance in controlling tumorigenesis and progression in various cancers. The role of gamma-interferon-inducible lysosomal thiol reductase (GILT) in tumor immunosurveillance has recently been studied in several malignant diseases, but its role in breast cancer remains to be elucidated. In the present study, we found GILT as a significant different expressed gene by cDNA microarray analysis. To further determine the role of GILT in breast cancer, we examined GILT expression in breast cancers as well as noncancerous breast tissues by immunohistochemistry and real-time PCR, and assessed its association with clinicopathologic characteristics and patient outcome. The absence of GILT expression increased significantly from 2.02% (2/99) in noncancerous breast tissues to 15.6% (34/218) in breast cancer tissues (P<0.001). In accordance with its proliferation inhibiting function, GILT expression was inversely correlated with Ki67 index (P<0.05). In addition, absence of GILT was positively correlated with adverse characteristics of breast cancers, such as histological type, tumor size, lymph nodes status, and pTNM stage (P<0.05). Consistently, breast cancers with reduced GILT expression had poorer disease-free survival (P<0.005). Moreover, significantly decreased expression of GILT was found in both primary and metastatic breast cancer cells, in contrast to normal epithelial cells. These findings indicate that GILT may act as a tumor suppressor in breast cancer, in line with its previously suggested role in anti-tumor immunity. Thus, GILT has the potential to be a novel independent prognostic factor in breast cancer and further studies are needed to illustrate the underlying mechanism of this relationship.