MilliporeSigma
ANKHD1, ankyrin repeat and KH domain containing 1, is overexpressed in acute leukemias and is associated with SHP2 in K562 cells.
Biochimica et biophysica acta (2006-09-08)
Fabíola Traina, Patricia M B Favaro, Samuel de Souza Medina, Adriana da Silva Santos Duarte, Sheila Maria Brochado Winnischofer, Fernando F Costa, Sara T O Saad
RÉSUMÉ
In the present study, increased levels of ANKHD1 mRNA and protein expression in leukemia cell lines are reported, as compared with normal hematopoietic cells. Furthermore, a higher expression of ANKHD1 mRNA was detected in primary acute leukemia samples than in normal hematopoietic cells (P=0.002). ANKHD1 was detected in the cytosolic and membrane fraction of cells and was co-immunoprecipitated with SHP2 in protein extracts of K562 and LNCaP cell lines. These findings suggest a role for ANKHD1 as a scaffolding protein that may be associated with the abnormal phenotype of leukemia cells.