MilliporeSigma
AKAP12/Gravin gene expression in colorectal cancer: clinical importance and review of the literature.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology (2013-09-26)
M Yildirim, D Suren, M Yildiz, A Sezgin Alikanoglu, R Eryilmaz, S Goktas, N Bulbuller, C Sezer
RÉSUMÉ
: Colorectal cancer (CRC) is a common and potentially lethal disease. A number of genetic aberrations is known to take place in colorectal carcinogenesis, which leads to progressive alteration of normal mechanisms controlling cell growth. A-kinase-anchoring protein 12 (AKAP12) plays a role in cell proliferation, angiogenesis and cytoskeletal remodeling. The purpose of this study was to demonstrate the role of the AKAP12 gene expression in CRC patients and to determine its relationship (if any) with prognosis. AKAP12 gene expression was investigated by immunohistochemistry. A total of 55 patients (63.6% males, 36.4% females) with histologically confirmed CRC were studied. Normal intestinal epithelium showed weak basal staining, dysplastic areas were stained mildly, whereas all of the cancer cells were stained completely with AKAP12. AKAP12 gene seems to play a role in colorectal carcinogenesis.