MilliporeSigma
The Rho kinase inhibitor Y-27632 increases erythrocyte deformability and low oxygen tension-induced ATP release.
American journal of physiology. Heart and circulatory physiology (2011-09-06)
K M Thuet, E A Bowles, M L Ellsworth, R S Sprague, A H Stephenson
RÉSUMÉ
Low oxygen (O(2)) tension and mechanical deformation are stimuli for ATP release from erythrocytes. It has been shown previously that rabbit erythrocytes made less deformable with diamide, a thiol cross-linking agent, release less ATP in response to low O(2) tension, suggesting a link between these two stimuli. In nonerythroid cells, activation of the Rho/Rho kinase signaling pathway has been reported to decrease cell deformability by altering Rho kinase-dependent cytoskeleton-protein interactions. We investigated the hypothesis that the Rho kinase inhibitor Y-27632 would increase erythrocyte deformability and thereby increase low O(2) tension-induced ATP release from erythrocytes. Here we show that Y-27632 (1 μM) increases erythrocyte deformability (5%) and increases low O(2) tension-induced ATP release (203%) from healthy human erythrocytes. In addition, we found that, when erythrocytes were made less deformable by incubation with diamide (100 μM), Y-27632 restored both deformability and low O(2) tension-induced ATP release to levels similar to those measured in the absence of diamide. These findings suggest that the Rho kinase inhibitor Y-27632 is able to reverse the diamide-induced decrease in erythrocyte deformability and rescue low O(2) tension-induced ATP release. These results further support a link between erythrocyte deformability and ATP release in response to low O(2) tension.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Mastoparan, Vespula lewisii, ≥97% (HPLC), powder