MilliporeSigma
Aminopeptidase activity in adult Schistosoma mansoni.
The Journal of parasitology (1983-04-01)
I M Cesari, C Auriault, A Capron
RÉSUMÉ
An aminopeptidase activity capable of hydrolysing leucine 4-nitroanilide and alanine 4-nitroanilide at pH 7.0 was detected in saponin-CaCl2 extracts and homogenates of adult Schistosoma mansoni. The extracts were also capable of acting on synthetic dipeptides at the same pH, preferentially hydrolysing peptide bonds following leucine, alanine, or proline N-terminal residues. Imide bonds were not hydrolysed. The hydrolysis of leucine 4-nitroanilide was apparently stimulated by thiols, strongly inhibited by 1 mM 4-chloromercuric benzene sulfonic acid, and partially inhibited by 1 mM 1,10-phenanthroline.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Leucine-p-nitroanilide, leucine aminopeptidase substrate