MilliporeSigma
  • Effects of breeding at the second oestrus or after post-weaning hormonal treatment with altrenogest on subsequent reproductive performance of primiparous sows.
Effects of breeding at the second oestrus or after post-weaning hormonal treatment with altrenogest on subsequent reproductive performance of primiparous sows.
Reproduction in domestic animals = Zuchthygiene (2011-02-18)
R F Werlang, L E Argenti, H C C Fries, M L Bernardi, I Wentz, F P Bortolozzo
RÉSUMÉ
This study investigated the effects of breeding at the second oestrus after weaning or after feeding an orally active progestagen (altrenogest) on the subsequent reproductive performance of primiparous sows. After 3 weeks of lactation, 663 weaned sows of two genotypes were allocated into three groups: G1--breeding at the first oestrus after weaning; G2--breeding at the second oestrus after weaning and G3--treatment with altrenogest for 5 days after weaning and breeding at the first oestrus after the end of the treatment. Body weight at breeding was lower in G1 and G3 than in G2 sows (p < 0.05). The interval to show oestrus was similar for G1 and G2 groups (p > 0.05) but higher (p < 0.05) than that observed in G3 group. Within genotype A, percentages of females in oestrus within 10 days were not different (p > 0.05) among groups, whereas in genotype B, more G1 and G2 sows (p < 0.05) showed oestrus than G3 sows. In both genotypes, lower farrowing rates were observed in G3 than in G1 and G2 sows (p < 0.05) and a greater litter size (p < 0.05) was observed in G2 sows. In genotype A, the number of total born piglets was similar for G1 and G3 groups (p > 0.05), whereas in genotype B, G1 sows had a greater litter size than G3 sows (p < 0.05). Body weight at weaning and at breeding was similar (p > 0.05) between farrowed and non-farrowed sows in all groups. Reproductive performance is not improved in primiparous sows treated with altrenogest during 5 days after weaning. The reproductive performance of genotype B sows is compromised in Control and Altrenogest-treated sows but not in those bred at the second oestrus after mating. Breeding at the second oestrus after weaning allows primiparous sows to gain weight between weaning and service, and increases their farrowing rate and subsequent litter size.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Altrenogest, VETRANAL®, analytical standard