MilliporeSigma
Inhibition of Toxoplasma gondii by indirubin and tryptanthrin analogs.
Antimicrobial agents and chemotherapy (2008-10-01)
Bogdana Krivogorsky, Peter Grundt, Robert Yolken, Lorraine Jones-Brando
RÉSUMÉ
New drugs are needed for treatment of Toxoplasma gondii infections. We tested derivatives of principles found in Isatis indigotica for in vitro efficacy against T. gondii infection. Indirubin-3'-oxime analogs showed modest micromolar activity, while tryptanthrin derivatives displayed 50% inhibitory doses in the low nanomolar range. Tryptanthrins have potential as anti-Toxoplasma infection therapeutics.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)