MilliporeSigma
[Biochemical aspects of growth-stimulating effects of steroid phytohormones on lupine plants].
Prikladnaia biokhimiia i mikrobiologiia (2007-07-11)
O L Kandelinskaia, A F Topunov, E R Grishchenko
RÉSUMÉ
Presowing treatment of seeds of lupine of various species and cultivars with brassinosteroids--homobrassinolide and epibrassinolide--caused an increase in protein content and a change in the proportion of some amino acids. Both hormones increased heterogeneity of high- and medium-molecular-weight nonhistone chromatin proteins but had no effect on the polypeptide profile of histones. Using epibrassinolide as an example, we showed that the brassinosteroid-induced increase in protein content in lupine seeds was due primarily to accumulation of the low-molecular-weight components of beta-conglutin. The content of some amino acids in both alpha- and beta-conglutins changed. These changes in protein metabolism correlated with an increase in the content of indoleacetic acid and a decrease in the content of abscisic acid.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
22(S),23(S)-Homobrassinolide