MilliporeSigma
Mechanisms for T-cell selective cytotoxicity of arabinosylguanine.
Blood (2003-05-17)
Carlos O Rodriguez, Christine M Stellrecht, Varsha Gandhi
RÉSUMÉ
Nelarabine, prodrug of arabinosylguanine (ara-G), has demonstrated T-lymphoblastic antileukemic activity in cell lines and in the clinic. To investigate the mechanism for lineage-specific toxicity, the effects of ara-G were compared in CEM (T-lymphoblast), Raji (B-lymphoblast), and ML-1 (myeloid) cell lines. CEM cells were the most sensitive to ara-G-induced apoptosis and accumulated the highest levels of ara-G triphosphate (ara-GTP). However, compared with myeloid and B-lineage cell lines, CEM cells incorporated fewer ara-G molecules-which were at internucleotide positions in all 3 cell lines- into DNA. Ara-G induced an S-phase arrest in both Raji and ML-1, while in CEM the S-phase cells decreased with a concomitant increase in the sub-G1 population. Within 3 hours of ara-G treatment, the levels of soluble Fas ligand (sFasL) in the medium increased significantly in CEM cultures. In parallel, an induction of FasL gene expression was observed by real-time reverse transcriptase-polymerase chain reaction (RT-PCR). Pretreatment of CEM cells with a Fas antagonistic antibody inhibited ara-G-mediated cell death. These results demonstrate that high ara-GTP accumulation in T cells results in an S phase-dependent apoptosis induced by ara-G incorporation into DNA, which may lead to a T cell-specific signal for the induction and liberation of sFasL. Subsequently, the sFasL induces an apoptotic response in neighboring non-S-phase cells. In contrast, myeloid and B cells accumulated lower levels of ara-GTP and arrested in S phase, blocking any apoptotic signaling.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ara-G hydrate, ≥98% (HPLC), solid