MilliporeSigma
Alteration and prognostic values of collagen gene expression in patients with gastric cancer under different treatments.
Pathology, research and practice (2020-02-02)
Xiaoyu Gao, Suhua Zhong, Yan Tong, Yushan Liang, Guofei Feng, Xiaoying Zhou, Zhe Zhang, Guangwu Huang
RÉSUMÉ
Collagen (COL) genes participate in tumor extracellular matrix (ECM)-receptor interactions and focal adhesion pathways, which play a crucial role in tumor invasion and metastasis. The prognostic value of COL genes has been shown for several malignancies. In the present study, we analyzed multiple microarray datasets using the Oncomine database to identify alterations of COL genes in gastric cancer (GC). Gene expression levels were analyzed by quantitative real-time polymerase chain reaction (qRT-PCR) and immunohistochemistry (IHC) in GC tissues and matched adjacent tissues. The prognostic value of differentially expressed COL genes in GC was evaluated by Kaplan-Meier survival analysis based on the complete mRNA transcriptomics data from The Cancer Genome Atlas (TCGA). We found that seven COL genes (COL1A2, COL4A1, COL4A2, COL6A1, COL6A2, COL6A3, and COL11A1) were elevated in GC. Among them, stepwise multivariate Cox regression was applied, and it was determined that COL4A1 and COL4A2 were signature and independent prognostic biomarkers in GC patients with obviously different overall survival (OS). High expression of COL4A1, COL4A2, COL6A1, COL6A2, and COL6A3 was correlated with poorer prognosis of GC patients treated by surgery only, while higher expression of COL4A1 and COL11A1 correlated with poorer survival of patients treated by 5-fluorouracil-based adjuvant therapy. Our results indicate that overexpression of COL genes might be utilized as novel prognostic markers for GC and assist with therapy selection.