MilliporeSigma
Influence of cultivar and technological conditions on the volatile profile of virgin pistachio oils.
Food chemistry (2019-12-22)
Rosa M Ojeda-Amador, Giuseppe Fregapane, María Desamparados Salvador
RÉSUMÉ
The main aim of this work was to characterize the volatile profile of virgin pistachio oils produced from eight cultivars (Aegina, Avdat, Kastel, Kerman, Larnaka, Mateur, Napoletana and Sirora), under different technological conditions (temperature, roasting, use of whole nuts, screw speed and nozzle diameter), and compare it with those of commercial pistachio oils. Terpenes (15.57-41.05 mg/kg), accounting for ~97% of total volatiles, were associated with appreciated sensory properties, with α-pinene as the main volatile (14.47-37.09 mg/kg). Other terpene compounds such as limonene (0.11-3.58 mg/kg), terpinolene (0.00-1.61 mg/kg), β-pinene (0.12-1.20 mg/kg) and α-terpineol (0.00-1.17 mg/kg) were quantified at lower concentrations. Acids, alcohols, aldehydes, esters and hydrocarbons only summed to ~3% of the total volatile compounds. The volatiles content greatly depended on the pistachio cultivar employed. The influence of extraction conditions was also very relevant; in particular, terpenes doubled (28.38-53.84 mg/kg) using whole pistachios for oil extraction, also being incremented by mild processing conditions. On the contrary, higher temperature or roasting decreased the terpene content (~50-25% respectively), and pyrazines appeared (up to 3.12 mg/kg).
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
o-Cymene, 98%