MilliporeSigma
Adsorption kinetics of the herbicide safeners, benoxacor and furilazole, to activated carbon and agricultural soils.
Journal of environmental sciences (China) (2020-01-02)
Saraswati Poudel Acharya, Jacob Johnson, Jennifer Weidhaas
RÉSUMÉ
Chloroacetamide herbicides, namely acetochlor and metolachlor, are common herbicides used on corn and soybean fields. Dichloroacetamide safeners, namely benoxacor and furilazole, are commonly used in formulations containing chloroacetamide herbicides. Extensive reports on adsorption of chloroacetamide herbicides are available, yet little information exists regarding adsorption potential of co-applied safeners. Herein, the adsorption and desorption characteristics of selected herbicide safeners to granular activated carbon (GAC) and in agricultural soils are reported. Further, soil column studies were performed to understand the leaching behaviour of the herbicide Dual II Magnum. Equilibrium sorption experiments of safeners to three agricultural soils and one GAC showed that adsorption was best fitted by the Freundlich isotherm. The Freundlich adsorption constant, Kf, for benoxacor and furilazole sorption onto three agricultural soils ranged from 0.1 to 0.27 and 0.1 to 0.13 (mg/g) × (mg/L)ˆ(1/n), respectively. The Kf for benoxacor and furilazole to GAC was 6.4 and 3.4 (mg/g) × (mg/L)ˆ(1/n), respectively, suggesting more favorable sorption of benoxacor to GAC than furilazole to GAC. The sorption to soils was reversible as almost 40%-90% of both safeners was desorbed from three soils. These results were validated in four replicated soil column studies, where S-metolachlor was shown to leach similarly to the safener benoxacor, originating from the herbicide formulation. The leaching of S-metolachlor and benoxacor was influenced by soil texture. Cumulatively, these results show that safeners will move through the environment to surface waters similarly to the active ingredients in herbicides, but may be removed during drinking water treatment via GAC.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
S-Metolachlor, PESTANAL®, analytical standard
Supelco
Benoxacor, PESTANAL®, analytical standard
Supelco
Furilazole, PESTANAL®, analytical standard