MilliporeSigma
Lipophilicity and bio-mimetic properties determination of phytoestrogens using ultra-high-performance liquid chromatography.
Biomedical chromatography : BMC (2019-04-14)
Kornelija Lasić, Ana Bokulić, Astrid Milić, Biljana Nigović, Ana Mornar
RÉSUMÉ
This paper presents lipophilicity and bio-mimetic property determination of 15 phytoestrogens, namely biochanin A, daidzein, formononetin, genistein, genistein-4,7-dimethylether, prunetin, 3,4,7-trihydroxyisoflavon, 4,6,7-trihydroxyisoflavon, 4,6,7-trimethoxyisoflavon, daidzin, genistin, ononin, sissotrin, coumestrol and coumestrol dimethylether. High-performance liquid chromatography with fast gradient elution and Caco-2 cell line were used to determine the physicochemical properties of selected phytoestrogens. Lipophilicity was determined on octadecyl-sylane stationary phase using pH 2.0 and pH 7.4 buffers. Immobilized artificial membrane chromatography was used for prediction of interaction with biological membranes. Protein binding was measured on human serum albumin and α-1-acid-glycoprotein (AGP) stationary phases. Caco-2 assay was used as a gold standard for assessing in vitro permeability. The obtained results differentiate phytoestrogens according to their structure where aglycones show significantly higher lipophilicity, immobilized artificial membrane partitioning, AGP binding and Caco-2 permeability compared with glucosides. However, human serum albumin binding was very high for all investigated compounds. Furthermore, a good correlation between experimentally obtained chromatographic parameters and in silico prediction was obtained for lipophilicity and human serum albumin binding, while the somewhat greater difference was obtained for AGP binding and Caco-2 permeability.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Amprenavir, ≥98% (HPLC)