MilliporeSigma
GSK3β and MCL-1 mediate cardiomyocyte apoptosis in response to high glucose.
Histochemistry and cell biology (2019-06-15)
Dongmei Su, Jing Zhao, Shanshan Hu, Lina Guan, Qian Li, Cuige Shi, Xu Ma, Jianjun Gou, Yunjun Zhou
RÉSUMÉ
Gestational diabetes mellitus is a risk factor for congenital heart defects. Our previous results indicated that a decrease in myocardial cells and an increase in apoptotic cells leads to heart defects under hyperglycemia, but much work remains to elucidate this important mechanism of myocardial cell apoptosis induced by high glucose (HG). In this study, we found that a decrease in GSK3β phosphorylation on Ser9 occurred concomitantly with HG-induced cardiomyocyte apoptosis and in the heart tissues of the offspring of diabetic rats in vitro and in vivo. Decreases in GSK3β (Ser9) phosphorylation in response to HG were remarkably restored after treatment with SC79, an activator of the Akt signaling pathway. SB216763, an effective inhibitor of the GSK3β signaling pathway, suppressed HG-induced apoptosis in cardiomyocytes. Further studies showed a decrease in the expression of the anti-apoptotic protein MCL-1 was associated with GSK3β-mediated apoptosis. MCL-1 overexpression partly inhibits HG-induced apoptosis in cardiomyocytes. Herein, this study revealed the roles of GSK3β and MCL-1 in modulating HG-induced cardiomyocyte apoptosis and maternal diabetes-induced abnormalities.