MilliporeSigma
Tumorigenesis and a DNA repair defect in mice with a truncating Brca2 mutation.
Nature genetics (1997-12-17)
F Connor, D Bertwistle, P J Mee, G M Ross, S Swift, E Grigorieva, V L Tybulewicz, A Ashworth
RÉSUMÉ
Germline mutation of the BRCA2 gene carries a high risk of developing breast cancer. To study the function of this gene, we generated a mutation in Brca2 in mice. Unlike other mutations in the Brca2 gene, which are lethal early in embryogenesis when homozygous, some of our homozygous mutant mice survive to adulthood. These animals have a wide range of defects, including small size, improper differentiation of tissues, absence of germ cells and the development of lethal thymic lymphomas. Fibroblasts cultured from BrcaZ-/-embryos have a defect in proliferation that may be mediated by over-expression of p53 and p21Waf1/CIP1. We show that Brca2 is required for efficient DNA repair, and our results suggest that loss of the p53 checkpoint may be essential for tumour progression triggered by mutations in BRCA2.