MilliporeSigma
  • Effects of reduced dietary energy and amino acid density on Pectoralis major myopathies in broiler chickens at 36 and 49 days of age1.
Effects of reduced dietary energy and amino acid density on Pectoralis major myopathies in broiler chickens at 36 and 49 days of age1.
Poultry science (2018-03-06)
K J Meloche, B I Fancher, D A Emmerson, S F Bilgili, W A Dozier
RÉSUMÉ
Two experiments (Exp) were conducted to determine if reductions in the incidence and severity of wooden breast (WB) and white striping (WS) may be obtained by reducing dietary nutrient density. In each Exp, Yield Plus × Ross 708 male broiler chicks were placed into 63 pens (22 birds/pen). All birds received an identical prestarter diet until 7 d of age, after which time each pen was randomly assigned to 1 of the following 7 dietary treatments (TRT) for the starter (8 to 14 d), grower (15 to 24 d), finisher 1 (Exp 1: 26 to 35 d; Exp 2: 26 to 42 d), and withdrawal (Exp 2: 43 to 48 d) phases: 1) 100% of primary breeder recommendations for digestible amino acid and metabolizable energy density throughout Exp; 2) 95% of TRT 1 until 14 d of age, then as TRT 1; 3) 95% of TRT 1 until 24 d of age, then as TRT 1; 4) 95% of TRT 1 throughout Exp; 5) 90% of TRT 1 until 14 d of age, then as TRT 1; 6) 90% of TRT 1 until 24 d of age, then as TRT 1; 7) 90% of TRT 1 throughout Exp. At 36 d (Exp 1) and 49 d (Exp 2), 18 birds per pen were processed and evaluated for WS and WB. In Exp 1, reduced dietary density in the starter phase (TRT 2 and TRT 5) resulted in increased (P ≤ 0.05) incidences of severe WB (32.9% and 34.7%) relative to TRT 1 (18.2%). In Exp 2, broilers assigned to TRT 7 had reduced (P < 0.01) incidences of severe WB (20.8%) and WS (42.3%) relative to the control (WB: 36.5%; WS: 64.5%). In both Exp, plasma creatine kinase and lactate dehydrogenase increased (P ≤ 0.05) with increasing scores for WB and WS. Reducing dietary nutrient density from 8 to 14 d may exacerbate fillet myopathies in broilers reared to 35 d of age. Although reducing dietary energy and amino acid density to 90% of recommendations from 1 to 48 d reduced the severity of myopathies, these reductions occurred with compromises in live performance. Altogether, these results indicated that concurrent manipulation of dietary amino acid and energy density is not a viable practical solution for breast myopathies.