MilliporeSigma
Jun turnover is controlled through JNK-dependent phosphorylation of the E3 ligase Itch.
Science (New York, N.Y.) (2004-09-11)
Min Gao, Tord Labuda, Ying Xia, Ewen Gallagher, Deyu Fang, Yun-Cai Liu, Michael Karin
RÉSUMÉ
The turnover of Jun proteins, like that of other transcription factors, is regulated through ubiquitin-dependent proteolysis. Usually, such processes are regulated by extracellular stimuli through phosphorylation of the target protein, which allows recognition by F box-containing E3 ubiquitin ligases. In the case of c-Jun and JunB, we found that extracellular stimuli also modulate protein turnover by regulating the activity of an E3 ligase by means of its phosphorylation. Activation of the Jun amino-terminal kinase (JNK) mitogen-activated protein kinase cascade after T cell stimulation accelerated degradation of c-Jun and JunB through phosphorylation-dependent activation of the E3 ligase Itch. This pathway modulates cytokine production by effector T cells.