MilliporeSigma
Distribution of S-100 protein and its subunits in bovine exocrine glands.
Histochemistry and cell biology (1997-07-01)
S Lauboeck, M Egerbacher
RÉSUMÉ
The distribution of S-100 protein and its alpha- and beta-subunits in bovine exocrine glands was studied by indirect immunohistochemistry. The entire spectrum of salivary glands, glands of the respiratory tract, intestinal glands, male and female genital glands, and skin glands was examined. S-100 and its beta-subunit were identified in most serous secretory cells of mixed salivary glands, although secretory acini in some serous glands remained unreactive for these antigens. Mucous cells were constantly negative; mucoid cells were positive in the lacrimal and Harderian gland. The alpha-subunit of S-100 protein was identified in serous cells but the staining reaction was faint. Subunits of S-100 showed a characteristic distribution along the excretory duct systems of compound glands: S-100 and the beta-subunit were present in intercalated duct epithelium, while striated duct epithelium stained for S100-alpha. Therefore, it is suggested that S100-alpha is related to resorption and secretion in striated ducts, while S100-beta may govern acinar exocytosis and probably regulates proliferation and differentiation of glandular cells. Differing staining intensities for S-100 and its subunits in secretory cells of exocrine glands most probably indicate functional differences with regard to secretory activity and the cell cycle.