MilliporeSigma
L-NAME improves doxycycline and ML-7 cardioprotection from oxidative stress.
Frontiers in bioscience (Landmark edition) (2017-09-21)
Iwona Bil-Lula, Anna Krzywonos-Zawadzka, Jolanta Sawicka, Dariusz Bialy, Magdalena Wawrzynska, Mieczyslaw Wozniak, Grzegorz Sawicki
RÉSUMÉ
Matrix metalloproteinase-2 (MMP-2) mediated degradation of myosin light chain 1 (MLC1) and troponin I (TnI) contributes to myocardial ischemia/reperfusion (I/R) injury. Modifications of MLC1 triggered by oxidative stress are mediated by myosin light chain kinase (MLCK), nitric oxide synthase (NOS), and MMP-2. Previous studies have shown that inhibiting both MLCK and MMP-2 protects against I/R injury. Here, we hypothesized that the addition of NOS inhibitor (L-NAME) at subprotective concentration to the mixture of subprotective concentrations of ML-7 and doxycycline (Doxy), will increase a synergistic cardioprotection of Doxy and ML-7 during I/R. Isolated rat hearts were subjected to global ischemia without or with administration of the mixture of inhibitors. Markers of I/R injury were measured in hearts and coronary effluents. Addition of L-NAME to the mixture of Doxy and ML-7 led to full recovery of heart contractility in comparison to combination of Doxy and ML-7. Improved heart contractility was associated with reduced degradation of TnI and MLC1. The combined administration of NOS, MMP-2 and MLCK inhibitors provides a novel strategy to protect heart from I/R injury.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ML-7, powder