MilliporeSigma
Trps1 is associated with the multidrug resistance of lung cancer cell by regulating MGMT gene expression.
Cancer medicine (2018-03-31)
Hongxiang Liu, Yi Liao, Meng Tang, Tao Wu, Deli Tan, Shixin Zhang, Haidong Wang
RÉSUMÉ
Multidrug resistance (MDR) often leads to chemotherapy failure of lung cancer and has been linking to the cellular expression of several DNA transcription- and repair-related genes such as Trps1 and MGMT. However, their roles in the formation of MDR are largely unknown. In this study, overexpression/knockdown, luciferase assay and ChIP assay were performed to study the relationship between Trps1 and MGMT, as well as their roles in MDR formation. Our results demonstrated that Trps1 and MGMT expression both increased in drug-resistant lung cancer cell line (H446/CDDP). Silencing of Trps1 resulted in downregulation of MGMT expression and decrease in the multidrug sensitivity of H446/CDDP cells, while Trps1 overexpression exhibited the opposite effects in H446 cells. Ectopic expression of MGMT had no effect on Trps1 expression, but enhanced the IC50 values of H446 cells or rescued the IC50 values of Trps1-silenced H446/CDDP cells in treatment of multidrug. Our data further showed that, mechanistically, Trps1 acted as a transcription activator that directly induced MGMT transcription by binding to the MGMT promoter. Taken together, we consider that upregulation of Trps1 induces MGMT transcription contributing to the formation of MDR in lung cancer cells. Our findings proved potential targets for reversing MDR in clinical chemotherapy of lung cancer.