MilliporeSigma
Système de purification d'eau Milli-Q® CLX Delivers up to 800 L/day clinical laboratory reagent water (CLRW). For feed water with high chlorine levels.
Système de purification d'eau Milli-Q® CLX 7040 (LC)
Système de purification d'eau Milli-Q® CLX Delivers up to 800 L/day clinical laboratory reagent water (CLRW). For feed water with high chlorine levels.
Synonyme(s):
Système de purification d'eau Milli-Q® CLX