MilliporeSigma
Adapter for Eppendorf® A-2-MTP Rotor holds 1 x 384-well PCR plate, pack of 2 ea
Adapter for Eppendorf® A-2-MTP Rotor
Adapter for Eppendorf® A-2-MTP Rotor holds 1 x 384-well PCR plate, pack of 2 ea