Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column
Products
Brand
Supelco (8)
Separation Technique
ID (mm)
Length (mm)
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
150487-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 100 Å, 3.5 μm particle size, L × I.D. 150 mm × 1.0 mm
Expand
View Pricing
150481-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 5 μm particle size, L × I.D. 150 mm × 0.3 mm
Expand
View Pricing
150480-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 3.5 μm particle size, L × I.D. 150 mm × 1.0 mm
Expand
View Pricing
150473-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 3.5 μm particle size, L × I.D. 50 mm × 0.3 mm
Expand
View Pricing
150479-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 3.5 μm particle size, L × I.D. 150 mm × 0.3 mm
Expand
View Pricing
150482-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 5 μm particle size, L × I.D. 150 mm × 1.0 mm
Expand
View Pricing
150476-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 200 Å, 3.5 μm particle size, L × I.D. 50 mm × 1.0 mm
Expand
View Pricing
150671-U
SeQuant
®
ZIC
®
-HILIC Stainless Steel Capillary HPLC Column,
pore size 100 Å, 3.5 μm particle size, L × I.D. 150 mm × 0.3 mm
Expand
View Pricing