
Mesocarbon microbeads

Synonym(s):
Graphite
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing