1A02620
USP
Sitagliptin (R)-Triazecine Analog

Synonym(s):
Sitagliptin (R)-Triazecine Analog, ((R)-10-(2,4,5-trifluorobenzyl)-3-(trifluoromethyl)-6,7,10,11-tetrahydro-[1,2,4]triazolo[3,4-c][1,4,7]triazecine-8,12(5H,9H)-dione)
About This Item
Recommended Products
grade
pharmaceutical analytical impurity (PAI)
Agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
-10 to -25°C
General description
Analysis Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service