Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1A00170

USP

Rosuvastatin Isoamyl Ester

new

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Rosuvastatin Isoamyl Ester, ((3S,5R,6E)-7-[4-(4-flurophenyl)-6-(1-methylethyl)-2-[N-methyl(methylsulphonyl)amino]-5-pyrimidinyI]-3,5-dihydroxy-6-heptanoic acid, methyl butyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38FN3O6S
CAS Number:
Molecular Weight:
551.67
MDL number:
UNSPSC Code:
12352200

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

-10 to -25°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service