Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1444285

USP

Mirtazapine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Desmethylmirtazapine, 1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, N-Demethylmirtazapine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3
CAS Number:
Molecular Weight:
251.33
Beilstein:
3976057
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C1CN2C(CN1)c3ccccc3Cc4cccnc24

InChI

1S/C16H17N3/c1-2-6-14-12(4-1)10-13-5-3-7-18-16(13)19-9-8-17-11-15(14)19/h1-7,15,17H,8-11H2

InChI key

FGLAMNFOHWVQOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid quantification of mirtazapine and desmethyl mirtazapine in human plasma by LC-ESI-MS/MS: Application to a bioequivalence study.
Kuchekar, S. R., M. L. Kundlik, and B. H. Zaware.
Journal of Saudi Chemical Society, 15.2, 145-153 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service