1225044
USP
Dofetilide Related Compound D
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Dofetilide Related Compound D, 4-{2-[(4-Aminophenethyl)(methyl)amino]ethoxy}aniline
About This Item
Recommended Products
grade
pharmaceutical primary standard
Agency
certified by the USP
API family
dofetilide
mol wt
285.39 (C17H23N3O)
packaging
pkg of 25 mg
manufacturer/tradename
USP 1225044
format
neat
storage temp.
2-8°C
Related Categories
General description
Application
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service