Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002353

Miconazole for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Miconazole nitrate salt, 1-(2,4-Dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl)imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14
MDL number:
UNSPSC Code:
12352200

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

ClC1=CC(Cl)=CC=C1C(OCC2=CC=C(Cl)C=C2Cl)CN3C=CN=C3.[O-][N+](O)=O

InChI

1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)

InChI key

MCCACAIVAXEFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Antifungal azole. Mode of action: Inhibits cytochrome P450-dependent 14α-demethylase, which is critical to ergosterol biosynthesis. The accumulated 14α-methylated sterols change the membrane structure of sensitive fungi, resulting in an altered cell membrane permeability. Also inhibits peroxidases, which results in accumulation of peroxide within the cell.
Miconazole interacts with 14-α demethylase, a cytochrome P-450 enzyme necessary for ergosterol biosynthesis. The inhibition of ergosterol results in increased cellular permeability causing leakage of cellular contents. Miconazole may also inhibit endogenous respiration, interact with phospholipids in the cell membrane, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and interfere with triglyceride and phospholipid biosynthesis.

Packaging

Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit EDQM reference substance catalogue.

Other Notes

EP issued certificate and SDS can be found here.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service