Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821805

TOYOPEARL MegaCap® II SP-550EC Bulk Media

bottle of 250 mL, phase sulphonyl, 200 μm particle size

Synonym(s):

TOYOPEARL MegaCap® II SP-550EC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 21805

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

sulphonyl phase

particle size

200 μm

operating pH

2-13

capacity

127.5 g/L±27.5 adsorption capacity (insulin)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl MegaCap® II SP-550EC resin is a 100 - 300 micron (EC) particle size strong cation exchanger useful for purification of molecules ranging in size from small peptides such as insulin to proteins up to 100kD. It is used for high throughput capture of feedstocks.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl MegaCap is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service