Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

819955

TOYOPEARL® SuperButyl-550C Bulk Media

bottle of 25 mL, phase C4 (butyl), 100 μm particle size

Synonym(s):

TOYOPEARL® SuperButyl-550 HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 19955

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

C4 (butyl) phase

particle size

100 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SuperButyl-550 resin has the second highest hydrophobicity of the HIC ligands offered by TBL. Its pore size is the smallest of our three butyl resins. Primary applications are for the separation of smaller proteins. Depending on the feedstock′s molecular size profile, it is also useful in negative or flow through chromatography. It can be used for capture and interresinte purification process steps

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service