Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814703

TOYOPEARL® Butyl-650C Bulk Media

phase C4 (butyl), bottle of 1 L, 100 μm particle size

Synonym(s):

TOYOPEARL® Butyl-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

description

Clean in place (CIP) (0.5 M NaOH or 0.1 M HCl)
Formulation (72% (v/v) slurry in 20% (v/v) ethanol)
Phase filter (Butyl)
Shipping solvent (20% (v/v) ethanol)

product line

TOYOPEARL®

feature

Molecular weight range (Excl. limit: 5,000,000 Da)

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14703

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

C4 (butyl) phase

particle size

100 μm

pore size

1000 Å

pH range

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

TOYOPEARL® HIC resins are hydrophobic interaction chromatography resins that offer the following advantages: strong affinity for water-soluble proteins, high recovery of mass and activity, high sample capacity up to 2-4 times that of gel media, fluctuating salt concentrations will not change the bed volume, mechanical stability to 7kg/cm squared (7 bar/100psi), stability at a wide pH range (2-12), clean in place with 0.5M NaOH, and autoclavable. The exclusion limit of 5x106 Da and large pore size, 1000Å, enables these packings to separate very large proteins by a hydrophobic interaction mechanism, without size exclusion effects.
TOYOPEARL® HIC resins for hydrophobic interaction chromatography offer these advantages:
  • Strong affinity for water-soluble proteins
  • High recovery of mass and activity
  • High sample capacity: up to 2-4 times that of gel media
  • Fluctuating salt concentrations will not change bed volume
  • Mechanical stability to 7kg/cm2 (7 bar/100psi)
  • Stable at wide pH range: 2-12
  • Clean in place with 0.5M NaOH
  • Autoclavable
     
TOYOPEARL media offer high yield recovery of proteins, using various aqueous eluants. The exclusion limit of 5x106 Da and large pore size, 1000Å, enables these packings to separate very large proteins by a hydrophobic interaction mechanism, without size exclusion effects. Packed as 100mL bottle.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Other Notes

Toyopearl Butyl-650 resin has the second highest hydrophobicity of the HIC ligands offered by TBL. Its pore size is the largest of our three butyl resins. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used in interresinte purification and polishing steps

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service