Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78717

Supelco

cis,cis-5,8-Tetradecanedienoyl-L-carnitine-(N-methyl-d3)

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[[(5Z,8Z)-1-oxo-5,8-tetradecadien-1-yl]oxy]-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D3H34NO4
Molecular Weight:
370.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

C[N+](C)(C([2H])([2H])[2H])C[C@H](OC(CCC/C=C\C/C=C\CCCCC)=O)CC([O-])=O

InChI

1S/C21H37NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4/h9-10,12-13,19H,5-8,11,14-18H2,1-4H3/b10-9-,13-12-/t19-/m1/s1/i2D3

InChI key

REUVFPFCXKWJQA-JEWRYSNVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service