Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69993

Supelco

2,3-Dimethylhydroquinone

analytical standard

Synonym(s):

1,4-Dihydroxy-2,3-dimethylbenzene, 2,3-Dimethyl-1,4-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2-1,4-(OH)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
636976
MDL number:

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

223-225 °C (lit.)

format

neat

SMILES string

Cc1c(C)c(O)ccc1O

InChI

1S/C8H10O2/c1-5-6(2)8(10)4-3-7(5)9/h3-4,9-10H,1-2H3

InChI key

BXJGUBZTZWCMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service