ZRB2718
Anti-CCR6/CD196 Antibody, clone 1K9 ZooMAb® Rabbit Monoclonal


recombinant, expressed in HEK 293 cells
Synonym(s):
C-C CKR-6, C-C chemokine receptor type 6, CKR-L3, Chemokine receptor-like 3, DRY6, G-protein coupled receptor 29, GPR-CY4, GPRCY4, LARC receptor
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1K9, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 42.49 kDa
observed mol wt ~57 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
Internal
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... CCR6(1235)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HuH-7 cell lysate.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected CCR6/CD196 in HuH-7 cell lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected CCR6/CD196 in human spleen tissue sections.
Flow Cytometry Analysis: 1 µg from a representative lot detected CCR6/CD196 in human peripheral blood mononuclear cells (PBMC).
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected CCR6/CD196 in HL-60 cells.
Affinity Binding Assay: A representative lot of this antibody bound CCR6/CD196 peptide with at least one hundred thousand-fold (100,000X) higher than with non-specific control peptide.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service