Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2669

Sigma-Aldrich

Anti-S1PR2 Antibody, clone 1C18 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Endothelial differentiation G-protein coupled receptor 5, S1P receptor 2, S1P receptor Edg-5, S1P2, Sphingosine 1-phosphate receptor 2, Sphingosine 1-phosphate receptor Edg-5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 38.87 kDa
observed mol wt ~48 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... S1PR2(9294)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C18 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects S1PR2. It targets an epitope within 25 amino acids from the N-terminal extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal, extracellular domain of human S1PR2.

Application

Quality Control Testing

Evaluated by Western Blotting in membrane extract from human cerebral cortex.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected S1PR2 in membrane extract from human cerebral cortex.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound S1PR2 peptide with at least one hundred thousand-fold (100,000X) higher than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected S1PR2 in human cerebral cortex tissue sections and human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Sphingosine 1-phosphate receptor 2 (UniProt: O95136; also known as S1P receptor 2, S1P2, Endothelial differentiation G-protein coupled receptor 5, Sphingosine 1-phosphate receptor Edg-5, S1P receptor Edg-5) is encoded by the S1PR2 (also known as EDG5) gene (Gene ID: 9294) in human. It is a ubiquitous G-protein coupled receptor for lysosphingolipid sphingosine 1-phosphate (S1P). S1P acts as an intercellular mediator by interacting with the endothelial differentiation gene (EDG)/S1P family of G protein-coupled receptors. It is coupled to Gi, G12/13, and Gq pathways to regulate a number of second messengers and effector molecules, including cAMP, PLC, MAPK, RhoA, and Rac. S1PR2 mediates the effects of S1P on cell motility and embryonic development, among other cellular processes. S1PR2 is widely expressed across various organs and cell types. It is prominently found in neuronal and vascular cells, macrophages, monocytes, and granulocytes. S1PR2 is a multi-pass membrane protein and primarily localized on the cell surface. S1P2 receptor is involved in S1P-induced platelet aggregation and Rho kinase activation. Activation of S1P2R in endothelial cells increases vascular permeability. It mediates immune cell trafficking, angiogenesis, and vascular function. S1PR2 activates downstream signaling pathways such as phosphoinositide 3-kinase (PI3K), Mitogen-activated protein kinase (MAPK), and Rho/Rho-associated coiled-coil containing kinases. S1PR2 is also known to be involved in the development of several diseases, including: Atherosclerotic disease, Alzheimer′s disease, Endotoxemia, and Multiple sclerosis. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service