Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2229

Sigma-Aldrich

Anti-Bid Antibody, clone 1G3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

BH3-interacting domain death agonist, p22 BID

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... BID(637)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G3 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Bid.

Immunogen

His-tagged, full-length, recombinant human Bid.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Bid in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:19,000 dilution from a representative lot detected Bid in lysate from MCF-7 cells and 1:1,000 dilution in lysates from C2C12, and PC12 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Bid in Human tonsil tissue sections.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected Bid in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

BH3-interacting domain death agonist (UniProt: P55957; also known as p22 BID, BID) is encoded by the BID gene (Gene ID: 637) in human. Bid is a BH3 domain containing member of the Bcl-2 family  with proapoptotic function. It is shown to be crucial for death receptor-mediated apoptosis in many cell systems where it counters the protective effects of Bcl-2. It links the death receptor signaling pathway to the mitochondrial signaling pathway mediated by Bcl-2 proteins. Full length Bid that is localized in the cytoplasm is cleaved by caspase-8 to generate a truncated form (tBid) that translocates to mitochondria. tBid then binds to mitochondrial Bak and induces the release of cytochrome c. TNFα-induced caspase-mediated cleavage of Bid produced p15, 13, and p11 fragments.  Caspase-8-mediated Bid processing of Bid bridges the extrinsic and intrinsic apoptosis pathways that provide a mechanism to amplify death signals.  The BH3 motif of Bid is localized in amino acids 86-100. The hydrophobic FACE® of the BH3 domain, which is exposed in other family members, is enclosed by the N-terminal of Bid and only exposed following proteolytic cleavage of Bid. A second functional domain in Bid known as the BH3B domain, which is localized in the N-terminus region, is reported to suppress the apoptogenic activity of the BH3 domain. The region between these two opposing domains is sensitive to proteolytic cleavage and contains cleavage sites for caspases and granzyme B. Four isoforms produced by Alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Renshaw, SA.,  et al. (2004). J. Biol. Chem. 279(4); 2846-2855; Esposti, MD. (2002). Apoptosis. 7; 433-440).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
FACE is a registered trademark of Glyko Inc.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service