Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

ZRB2210

Sigma-Aldrich

Anti-SETD5 Antibody, clone 3B22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Histone-lysine N-methyltransferase SETD5, SET domain-containing protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 3B22
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 157.5 kDa
observed mol wt ~160 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SETD5(55209)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3B22 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects SET domain-containing protein 5 (SETD5). It targets an epitope within the internal region.

Immunogen

His-tagged recombinant fragment corresponding to 206 amino acids from the internal region of human SET domain-containing protein 5 (SETD5).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SETD5 in HeLa cell nuclear extract.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SETD5 in Raji cell nuclear extract.

Affinity Binding Assay: A representative lot of this antibody bound recombinant fragment of Human SETD5 with a KD of 9.7 x 10-9 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SETD5 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Histone-lysine N-methyltransferase SETD5 (UniProt: Q9C0A6; also known as SET domain-containing protein 5, SETD5) is encoded by the SETD5 (also known as KIAA1757) gene (Gene ID: 55209) in human. SETD5 is shown to be essential for mammalian development and is required for regulating germ cell-associated genes in embryonic stem cells. It localizes to active transcribed genes and its SET domain is localized to amino acids 269-390. It interacts with Tbl1xr1 and Ctr9, which are components of 2 different transcriptional regulatory complexes (NcoR1 corepressor complex and Paf1 complex) in murine embryonic stem cells. SETD5-deficient embryonic stem cells display impaired cellular proliferation, increased apoptosis, defective cell cycle progression, a diminished ability to differentiate into cardiomyocytes, and greatly perturbed gene expression. SETD5-deficient mice embryos exhibit severe defects in neural tube formation, somitogenesis, and cardiac development, have aberrant vasculogenesis in embryos, yolk sacs, and placentas. They die between embryonic day 10.5 and 11.5. SETD5 may act by mediating trimethylation of lysine 36 of histone H3 (H3K36me3), which is essential to allow on-time RNA elongation dynamics. It can also monomethylates lysine 9 of histone H3 (H3K9me1) in vitro. Three isoforms of SETD5 have been described that are produced by alternative splicing. Mutation in SETD5 gene has been linked to intellectual disability. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Yu, SE., et al. (2017). Cell. Biochem. Funct. 35(5); 247-253; Osipovich, AB., et al. (2016). Development 143(24); 4595-4607).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service